Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004444.1 | - | 8 | 1255 | ||
AK076500.1 | - | 8 | 1242 | |||
AK151245.1 | - | 7 | 1061 | |||
AK158058.1 | - | 8 | 1317 | |||
AK205223.1 | - | 1 | 140 | |||
AK206061.1 | - | 1 | 140 | |||
BC033921.1 | - | 8 | 1227 | 5068809 |