Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hav1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028864.2 | NP_083140.1 | 9 | 2901 | ||
GenBank mRNA | AK004770.1 | - | 9 | 2902 | ||
AK080334.1 | - | 9 | 2886 | |||
AK169402.1 | - | 9 | 2546 | |||
AK172379.1 | - | 9 | 2902 | |||
BC029090.1 | - | 9 | 2463 | 2651558 |