Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185060.1 | NP_001171989.1 | 5 | 859 | ||
NM_001185061.1 | NP_001171990.1 | 4 | 1034 | |||
NM_001185062.1 | NP_001171991.1 | 4 | 806 | |||
NM_198098.2 | NP_932766.1 | 4 | 1179 | |||
GenBank mRNA | AK298091.1 | - | 5 | 1081 | ||
AK298313.1 | - | 5 | 940 | |||
AK300995.1 | - | 5 | 991 | |||
AK301351.1 | - | 4 | 787 | |||
AK309608.1 | - | 5 | 859 | |||
AK309691.1 | - | 4 | 878 | |||
AY312580.1 | - | 3 | 288 | |||
BC022486.1 | - | 4 | 1126 | 4799283 | ||
M77829.1 | - | 4 | 1107 | |||
S73482.1 | - | 4 | 1107 | |||
U41517.1 | - | 4 | 1102 | |||
U41518.1 | - | 4 | 720 |