Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm4070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243039.1 | NP_001229968.1 | 4 | 486 | ||
NM_001243040.1 | NP_001229969.1 | 2 | 292 | |||
GenBank mRNA | AK036418.1 | - | 4 | 476 | ||
AY167972.1 | - | 4 | 475 | |||
AY167973.1 | - | 2 | 265 | |||
AY167974.1 | - | 4 | 459 | |||
BY199517.1 | - | 3 | 347 |
Gene Symbol: Gvin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039160.2 | NP_001034249.2 | 2 | 295 | ||
NM_029000.3 | NP_083276.2 | 4 | 489 | |||
GenBank mRNA | AK154796.1 | - | 4 | 478 |