Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029001.5 | NP_083277.3 | 4 | 453 | ||
GenBank mRNA | AK018616.1 | - | 4 | 428 | ||
AK037162.1 | - | 4 | 425 | |||
AK049118.1 | - | 4 | 453 | |||
AK050441.1 | - | 4 | 402 | |||
AK137668.1 | - | 4 | 438 | |||
BC005602.1 | - | 4 | 453 | 3483017 |