Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290689.1 | NP_001277618.1 | 2 | 267 | ||
NM_029090.3 | NP_083366.1 | 3 | 383 | |||
GenBank mRNA | AI448494.1 | - | 2 | 252 | ||
AK004750.1 | - | 3 | 276 | |||
AK039910.1 | - | 4 | 789 | |||
AK088736.1 | - | 3 | 301 | |||
BC004837.1 | - | 3 | 319 | 2654387 | ||
BY255312.1 | - | 3 | 301 |