Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029103.3 | NP_083379.2 | 1, 2 | 121 | ||
GenBank mRNA | AB009291.1 | - | 1, 2 | 95 | ||
AK014338.1 | - | 1, 2 | 99 | |||
AK131997.1 | - | 1, 2 | 122 | |||
AK165646.1 | - | 1, 2 | 97 | |||
AK167580.1 | - | 1, 2 | 122 | |||
AK176996.1 | - | 1, 2 | 15 | |||
AK186239.1 | - | 1, 2 | 15 | |||
BC022761.1 | - | 1, 2 | 129 | |||
BC038901.1 | - | 1, 2 | 109 |