Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynlrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029297.1 | NP_083573.1 | 2, 3 | 119 | ||
GenBank mRNA | AK005789.1 | - | 2, 3 | 119 | ||
BC048623.1 | - | 2, 3 | 160 | 6743790 |