Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029020.2 | NP_083296.2 | 11 | 1692 | ||
GenBank mRNA | AK014319.1 | - | 11 | 1423 | ||
AK030624.1 | - | 11 | 1419 | |||
AK038760.1 | - | 11 | 1420 | |||
AK049152.1 | - | 11 | 1436 | |||
AK076315.1 | - | 11 | 1443 | |||
AK166704.1 | - | 11 | 1426 | |||
AK189429.1 | - | 1 | 33 | |||
AK196341.1 | - | 1 | 98 | |||
AK205793.1 | - | 1 | 46 | |||
BC015301.1 | - | 11 | 1692 | 4237894 | ||
BC050242.1 | - | 1 | 407 |