Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddit4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029083.2 | NP_083359.1 | 3 | 892 | ||
GenBank mRNA | AK017926.1 | - | 3 | 892 | ||
AK081046.1 | - | 3 | 956 | |||
AY260552.1 | - | 2 | 815 | |||
BC131992.1 | - | 3 | 891 | 40130269 | ||
BC132645.1 | - | 3 | 891 | 40130922 | ||
BC145370.1 | - | 3 | 891 | 9053928 |