Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029091.2 | NP_083367.1 | 2 | 262 | ||
GenBank mRNA | AK004774.1 | - | 2 | 263 | ||
AK170793.1 | - | 2 | 246 | |||
BC005746.1 | - | 2 | 284 | 3599898 |