Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hhatl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029095.2 | NP_083371.2 | 6 | 765 | ||
GenBank mRNA | AK003605.1 | - | 6 | 642 | ||
AK157924.1 | - | 6 | 563 | |||
BC024464.1 | - | 6 | 621 | 4975643 |