Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028994.2 | NP_083270.1 | 10 | 2987 | ||
GenBank mRNA | AK030501.1 | - | 10 | 2988 | ||
AK031612.1 | - | 10 | 2832 | |||
AK142561.1 | - | 4 | 1739 | |||
AK165094.1 | - | 11 | 2565 | |||
BC028326.1 | - | 6 | 1487 |