Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029020.2 | NP_083296.2 | 16 | 2878 | ||
GenBank mRNA | AK014319.1 | - | 16 | 2609 | ||
AK030624.1 | - | 16 | 2605 | |||
AK038760.1 | - | 16 | 2606 | |||
AK049152.1 | - | 16 | 2622 | |||
AK076315.1 | - | 16 | 2631 | |||
AK166704.1 | - | 16 | 2612 | |||
AK170334.1 | - | 4 | 898 | |||
AK186503.1 | - | 1 | 315 | |||
AK212391.1 | - | 1 | 228 | |||
AK218490.1 | - | 1 | 228 | |||
BC015301.1 | - | 16 | 2878 | 4237894 | ||
BC050242.1 | - | 4 | 2272 |