Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029091.2 | NP_083367.1 | 16 | 2114 | ||
GenBank mRNA | AK004774.1 | - | 16 | 2115 | ||
AK170793.1 | - | 16 | 2098 | |||
AK189481.1 | - | 1 | 83 |