Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000416.2 | NP_000407.1 | 2 | 276 | ||
XM_006715470.2 | XP_006715533.1 | 3 | 637 | |||
XM_006715471.2 | XP_006715534.1 | 2 | 526 | |||
XM_011535793.2 | XP_011534095.1 | 3 | 1308 | |||
XM_011535794.1 | XP_011534096.1 | 3 | 253 | |||
XM_017010827.1 | XP_016866316.1 | 3 | 347 | |||
GenBank mRNA | AF056979.1 | - | 2 | 194 | ||
AK222803.1 | - | 2 | 231 | |||
AK294252.1 | - | 3 | 319 | |||
AK297052.1 | - | 2 | 152 | |||
AK312674.1 | - | 2 | 230 | |||
BC005333.1 | - | 2 | 255 | 3950528 | ||
BT006814.1 | - | 2 | 173 | |||
DC349115.1 | - | 2 | 276 | |||
EF535103.1 | - | 2 | 216 | |||
J03143.1 | - | 2 | 221 | |||
KU178144.1 | - | 2 | 173 | |||
KU178145.1 | - | 2 | 173 |