Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029186.2 | NP_083462.2 | 9 | 2234 | ||
GenBank mRNA | AK015988.1 | - | 6 | 1588 | ||
AK031554.1 | - | 9 | 2180 | |||
AK052797.1 | - | 5 | 1220 | |||
AK151797.1 | - | 9 | 2209 | |||
AK161305.1 | - | 9 | 2190 | |||
AK170247.1 | - | 2180 | ||||
BC026385.1 | - | 6 | 2190 | |||
BC031169.1 | - | 8 | 2007 | |||
BC049142.1 | - | 6 | 1520 | |||
BC058746.1 | - | 9 | 2180 | 6399588 |