Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029402.3 | NP_083678.2 | 6 | 646 | ||
GenBank mRNA | AK004875.2 | - | 6 | 602 | ||
AK016520.1 | - | 6 | 616 | |||
AK144693.1 | - | 6 | 619 | |||
AK160597.1 | - | 6 | 631 | |||
AK220287.1 | - | 6 | 621 | |||
BC026779.1 | - | 6 | 547 | 4018298 | ||
BC027428.1 | - | 6 | 609 | 3983461 |