Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pibf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029320.3 | NP_083596.2 | 2 | 849 | ||
GenBank mRNA | AK015781.1 | - | 2 | 316 | ||
AK017107.1 | - | 2 | 850 | |||
AK030385.1 | - | 2 | 330 | |||
AK045133.1 | - | 2 | 344 | |||
AK050974.1 | - | 1 | 1216 | |||
BC129811.1 | - | 2 | 326 | 40062382 | ||
BC129812.1 | - | 2 | 326 | 40062383 |