Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029555.2 | NP_083831.1 | 1, 2 | 167 | ||
GenBank mRNA | AK002661.1 | - | 1, 2 | 165 | ||
AY279096.1 | - | 1, 2 | 80 | |||
BC038151.1 | - | 1, 2 | 94 | |||
BC051507.1 | - | 1, 2 | 131 | |||
BC058163.1 | - | 1, 2 | 100 | 5100572 |