Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029565.3 | NP_083841.4 | 6 | 763 | ||
GenBank mRNA | AK003252.1 | - | 6 | 677 | ||
AK014184.1 | - | 6 | 549 | |||
AK050162.1 | - | 6 | 681 | |||
AK145644.1 | - | 6 | 684 | |||
AK159761.1 | - | 6 | 691 | |||
AK166772.1 | - | 6 | 678 | |||
AK167058.1 | - | 6 | 677 | |||
AK177231.1 | - | 2 | 96 | |||
AX775905.1 | - | 6 | 698 | |||
BC014732.1 | - | 6 | 667 | 4235248 | ||
BC018379.1 | - | 6 | 685 | 4242583 | ||
BC045145.1 | - | 6 | 701 | 6475115 | ||
BC058273.1 | - | 6 | 678 | 6494379 |