Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029582.2 | NP_083858.1 | 8 | 1837 | ||
NM_134105.2 | NP_598866.1 | 1 | 80 | |||
GenBank mRNA | AK013044.1 | - | 8 | 1885 | ||
AK139620.1 | - | 8 | 1920 | |||
AK155844.1 | - | 8 | 1891 | |||
AK156154.1 | - | 8 | 1878 | |||
AK157288.1 | - | 8 | 1811 | |||
AK157948.1 | - | 8 | 1917 | |||
AK159484.1 | - | 8 | 1894 | |||
AK170352.1 | - | 7 | 1765 | |||
AK177768.1 | - | 1 | 156 | |||
AK206713.1 | - | 1 | 157 | |||
AK216234.1 | - | 1 | 155 | |||
BC019564.1 | - | 1 | 105 | |||
BC029643.1 | - | 8 | 1837 | 4010810 |