Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029595.3 | NP_083871.3 | 1 | 226 | ||
GenBank mRNA | AF307146.1 | - | 1 | 206 | ||
AF307147.1 | - | 1 | 206 | |||
AK006258.1 | - | 1 | 251 | |||
BC107334.1 | - | 1 | 226 | 40054986 | ||
BC107335.1 | - | 1 | 226 | 40054988 |