Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029600.3 | NP_083876.3 | 31 | 4549 | ||
GenBank mRNA | AF534127.1 | - | 31 | 4552 | ||
AF534128.1 | - | 31 | 4477 | |||
AF534129.1 | - | 30 | 4630 | |||
AK006128.1 | - | 4 | 533 | |||
AK149551.1 | - | 31 | 4549 | |||
AK155664.1 | - | 31 | 4527 | |||
AK156615.1 | - | 31 | 4779 | |||
AK172420.1 | - | 31 | 4779 | |||
AY795569.1 | - | 31 | 4549 | |||
AY841885.1 | - | 31 | 4530 | |||
BC046560.1 | - | 28 | 4056 | |||
BC048825.1 | - | 31 | 4481 | |||
BC058185.1 | - | 8 | 998 | |||
BC150788.1 | - | 31 | 4543 | 9087699 |