Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAFAH1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000430.3 | NP_000421.1 | 3 | 675 | ||
XM_011523901.1 | XP_011522203.1 | 4 | 724 | |||
XM_011523902.2 | XP_011522204.1 | 4 | 556 | |||
XM_011523903.2 | XP_011522205.1 | 4 | 474 | |||
XM_017024699.1 | XP_016880188.1 | 3 | 425 | |||
XM_017024700.1 | XP_016880189.1 | 3 | 502 | |||
XM_017024701.1 | XP_016880190.1 | 3 | 418 | |||
GenBank mRNA | AF208835.1 | - | 2 | 107 | ||
AF208836.1 | - | 2 | 107 | |||
AF208837.1 | - | 2 | 107 | |||
AF208838.1 | - | 2 | 107 | |||
AF400434.1 | - | 2 | 300 | |||
AK313078.1 | - | 3 | 329 | |||
BC064638.1 | - | 3 | 317 | 5786560 | ||
L13385.1 | - | 3 | 324 | |||
L13386.1 | - | 3 | 560 |