Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029410.3 | NP_083686.1 | 5 | 535 | ||
GenBank mRNA | AK013411.1 | - | 4 | 600 | ||
AK017362.1 | - | 5 | 536 | |||
BC039807.1 | - | 687 | 1529147 |