Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193309.1 | NP_001180238.1 | 3 | 517 | ||
NM_029413.4 | NP_083689.2 | 3 | 517 | |||
GenBank mRNA | AK017472.1 | - | 3 | 486 | ||
AK032807.1 | - | 3 | 518 | |||
AK048519.1 | - | 3 | 486 | |||
AK053895.1 | - | 3 | 489 | |||
AK164017.1 | - | 3 | 485 | |||
BC098483.1 | - | 3 | 472 | 30022418 | ||
BC132153.1 | - | 3 | 371 | 40130430 | ||
BC132497.1 | - | 3 | 371 | 40130774 |