Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000436.3 | NP_000427.1 | 1, 2 | 417 | ||
XM_017009494.1 | XP_016864983.1 | 1, 2 | 298 | |||
GenBank mRNA | AK312327.1 | - | 1, 2 | 113 | ||
BC009001.1 | - | 1, 2 | 141 | 4148643 | ||
BE244429.1 | - | 1, 2 | 232 | |||
CR457049.1 | - | 1, 2 | 86 | |||
U62961.1 | - | 1, 2 | 184 |