Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029600.3 | NP_083876.3 | 31 | 4597 | ||
GenBank mRNA | AF534127.1 | - | 31 | 4600 | ||
AF534128.1 | - | 31 | 4525 | |||
AF534129.1 | - | 30 | 4678 | |||
AK006128.1 | - | 4 | 581 | |||
AK149551.1 | - | 31 | 4597 | |||
AK155664.1 | - | 31 | 4575 | |||
AK156615.1 | - | 31 | 4827 | |||
AK172420.1 | - | 31 | 4827 | |||
AY795569.1 | - | 31 | 4597 | |||
AY841885.1 | - | 31 | 4578 | |||
BC046560.1 | - | 28 | 4104 | |||
BC048825.1 | - | 31 | 4529 | |||
BC058185.1 | - | 8 | 1046 | |||
BC150788.1 | - | 31 | 4591 | 9087699 |