Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029624.4 | NP_083900.3 | 7 | 1024 | ||
GenBank mRNA | AK010312.1 | - | 7 | 995 | ||
AK041659.1 | - | 5 | 691 | |||
AK154622.1 | - | 7 | 1021 | |||
AK169983.1 | - | 7 | 1022 | |||
AK172374.1 | - | 6, 7 | 897 | |||
AK177501.1 | - | 2 | 156 | |||
AK183502.1 | - | 2 | 156 | |||
BC020104.1 | - | 7 | 1031 | 3963699 |