Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pih1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278207.1 | NP_001265136.1 | 4 | 317 | ||
NM_001285904.1 | NP_001272833.1 | 4 | 309 | |||
NM_029406.4 | NP_083682.1 | 4 | 437 | |||
GenBank mRNA | AK004341.1 | - | 4 | 318 | ||
AK016797.1 | - | 4 | 343 | |||
AK089003.1 | - | 4 | 386 | |||
AK146039.1 | - | 4 | 375 | |||
AK152695.1 | - | 4 | 306 | |||
AK177389.1 | - | 3 | 183 | |||
BC068254.1 | - | 4 | 326 | 30013096 |