Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc10a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029415.2 | NP_083691.1 | 5 | 943 | ||
GenBank mRNA | AJ583504.1 | - | 5 | 772 | ||
AK018423.1 | - | 5 | 944 | |||
BC127608.1 | - | 5 | 836 | 40058121 | ||
BC139343.1 | - | 5 | 835 | 8862365 | ||
BC139346.1 | - | 5 | 835 | 8862368 |