Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000446.5 | NP_000437.3 | 2 | 204 | ||
GenBank mRNA | AK314027.1 | - | 2 | 130 | ||
BC074719.2 | - | 2 | 136 | 30915169 | ||
BJ993711.1 | - | 2 | 204 | |||
CB122285.1 | - | 2 | 164 | |||
D84371.1 | - | 2 | 161 | |||
M63012.1 | - | 2 | 116 | |||
M63013.1 | - | 2 | 55 | |||
M63014.1 | - | 1 | 21 | |||
S64615.1 | - | 2 | 116 | |||
U53784.1 | - | 2 | 161 | |||
Z70723.1 | - | 2 | 161 |