Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029457.3 | NP_083733.1 | 11 | 1185 | ||
GenBank mRNA | AF368904.1 | - | 11 | 1231 | ||
AK015987.1 | - | 11 | 1164 | |||
AK031030.1 | - | 11 | 1170 | |||
AY188288.1 | - | 10 | 957 | |||
BC031652.1 | - | 11 | 1160 | 3483046 | ||
BC040805.1 | - | 3 | 1129 | |||
BC042514.1 | - | 3 | 1129 |