Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029478.3 | NP_083754.2 | 12 | 1323 | ||
GenBank mRNA | AB047550.1 | - | 12 | 1335 | ||
AB076609.1 | - | 11 | 1214 | |||
AK043091.1 | - | 12 | 1324 | |||
AK049559.1 | - | 12 | 1316 | |||
AK077443.1 | - | 12 | 1323 | |||
AK154616.1 | - | 1 | 1620 | |||
AK159468.1 | - | 12 | 1338 | |||
AK167173.1 | - | 12 | 1331 | |||
AK167202.1 | - | 12 | 1331 | |||
AK211254.1 | - | 4 | 385 | |||
AK217200.1 | - | 1 | 151 | |||
AX775963.1 | - | 12 | 1293 | |||
BC004013.1 | - | 12 | 1317 | 3493738 |