Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Keg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029550.4 | NP_083826.1 | 6 | 1196 | ||
GenBank mRNA | AB028071.1 | - | 6 | 1172 | ||
AK002360.1 | - | 6 | 1194 | |||
AK165515.1 | - | 6 | 1194 | |||
BC010803.1 | - | 6 | 1186 | 4238654 |