Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029573.2 | NP_083849.1 | 5 | 465 | ||
GenBank mRNA | AK003393.1 | - | 2 | 358 | ||
AK010065.1 | - | 5 | 489 | |||
AK032787.1 | - | 5 | 489 | |||
AK047951.1 | - | 5 | 487 | |||
AK117103.1 | - | 5 | 501 | |||
AK146717.1 | - | 2 | 329 | |||
AK150618.1 | - | 5 | 496 | |||
AK151304.1 | - | 5 | 490 | |||
AK152353.1 | - | 5 | 499 | |||
AK153459.1 | - | 5 | 490 | |||
AK158646.1 | - | 5 | 643 | |||
AK159051.1 | - | 5 | 562 | |||
AK159666.1 | - | 5 | 496 | |||
AK160054.1 | - | 5 | 496 | |||
AK164038.1 | - | 6 | 591 | |||
AK167822.1 | - | 5 | 495 | |||
AK168034.1 | - | 5 | 487 | |||
AK168049.1 | - | 5 | 490 | |||
AK168149.1 | - | 5 | 496 | |||
AK169152.1 | - | 5 | 463 | |||
BC034273.1 | - | 5 | 455 | 4166193 | ||
BC049956.1 | - | 5 | 465 | 4212315 |