Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcolce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029620.2 | NP_083896.1 | 9 | 1804 | ||
GenBank mRNA | AK010249.1 | - | 9 | 1805 | ||
AK132395.1 | - | 1694 | ||||
BC051174.1 | - | 9 | 1677 | 6310294 |