Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000326.4 | NP_000317.1 | 4, 5 | 529 | ||
XM_011521870.2 | XP_011520172.1 | 4, 5 | 1958 | |||
XM_017022460.1 | XP_016877949.1 | 1, 2 | 368 | |||
GenBank mRNA | AK312457.1 | - | 4, 5 | 417 | ||
BC004199.2 | - | 4, 5 | 431 | 3604048 | ||
BM921294.1 | - | 4, 5 | 598 | |||
J04213.1 | - | 3, 4 | 332 |