Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029669.4 | NP_083945.1 | 2 | 216 | ||
GenBank mRNA | AK012521.1 | - | 2 | 169 | ||
AK030483.1 | - | 2 | 194 |