Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253681.1 | NP_001240610.1 | 15 | 1845 | ||
NM_001253686.1 | NP_001240615.1 | 14 | 1830 | |||
NM_001253687.1 | NP_001240616.1 | 13 | 1734 | |||
NM_001253688.1 | NP_001240617.1 | 13 | 1644 | |||
NM_001253689.1 | NP_001240618.1 | 13 | 1615 | |||
NM_029673.3 | NP_083949.2 | 15 | 1878 | |||
GenBank mRNA | AK006977.1 | - | 3 | 223 | ||
AK033126.1 | - | 13 | 1645 | |||
AK038129.1 | - | 15 | 1868 | |||
AK049189.1 | - | 15 | 1789 | |||
AK146640.1 | - | 13 | 1607 | |||
AK152588.1 | - | 14 | 1776 | |||
AK159909.1 | - | 14 | 1781 | |||
AK163491.1 | - | 15 | 1753 | |||
AK169365.1 | - | 13 | 1547 | |||
BC025034.1 | - | 3 | 623 | |||
BC055840.1 | - | 3 | 307 | |||
BC061010.1 | - | 4 | 349 | |||
BC080790.1 | - | 2 | 195 | |||
BC108356.1 | - | 7 | 776 | |||
BC145704.1 | - | 2 | 158 | |||
BC145706.1 | - | 2 | 158 |