Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcun1d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029775.2 | NP_084051.1 | 2 | 368 | ||
GenBank mRNA | AK012764.1 | - | 2 | 381 | ||
AK013933.1 | - | 2 | 386 | |||
AK014737.1 | - | 2 | 410 | |||
AK134862.1 | - | 415 | ||||
BC020048.1 | - | 2 | 404 | |||
BC020089.1 | - | 2 | 368 | 4013755 |