Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029787.2 | NP_084063.1 | 2, 3 | 234 | ||
GenBank mRNA | AF332059.1 | - | 2, 3 | 158 | ||
AF332060.1 | - | 2, 3 | 158 | |||
AK002640.1 | - | 2, 3 | 206 | |||
AK010858.1 | - | 2, 3 | 211 | |||
AK128917.1 | - | 2, 3 | 228 | |||
AK202011.1 | - | 1, 2 | 64 | |||
AK208840.1 | - | 1, 2 | 64 | |||
BC004760.1 | - | 2, 3 | 241 | 3154531 | ||
BC032013.1 | - | 2, 3 | 216 | 4238275 | ||
BC043074.1 | - | 2, 3 | 234 | 6404746 |