Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bicd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039179.2 | NP_001034268.1 | 2 | 382 | ||
NM_001039180.2 | NP_001034269.1 | 3 | 278 | |||
NM_029791.4 | NP_084067.1 | 2 | 382 | |||
GenBank mRNA | AJ250106.1 | - | 2 | 341 | ||
AK002683.1 | - | 2 | 64 | |||
AK122348.1 | - | 2 | 355 | |||
AK156407.1 | - | 2 | 337 | |||
AK171300.1 | - | 3 | 236 | |||
BC032198.1 | - | 2 | 339 | 5324113 |