Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029793.2 | NP_084069.1 | 4, 5 | 559 | ||
GenBank mRNA | AK002954.1 | - | 4, 5 | 503 | ||
AK008972.1 | - | 4, 5 | 459 | |||
AK028516.1 | - | 4, 5 | 478 | |||
AK044987.1 | - | 4, 5 | 507 | |||
BC024637.1 | - | 4, 5 | 481 | |||
BC043090.1 | - | 4, 5 | 424 | 6418353 |