Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg16l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205391.1 | NP_001192320.1 | 3 | 463 | ||
NM_001205392.1 | NP_001192321.1 | 3 | 463 | |||
NM_029846.4 | NP_084122.2 | 3 | 463 | |||
GenBank mRNA | AB087879.1 | - | 3 | 274 | ||
AB087880.1 | - | 3 | 274 | |||
AB087881.1 | - | 3 | 274 | |||
AB476646.1 | - | 3 | 274 | |||
AK005181.1 | - | 3 | 388 | |||
AK030983.1 | - | 3 | 404 | |||
AK048873.1 | - | 3 | 382 | |||
AK131120.1 | - | 3 | 362 | |||
AK134333.1 | - | 3 | 419 | |||
AK145673.1 | - | 3 | 401 | |||
AK155358.1 | - | 3 | 399 | |||
AK163246.1 | - | 3 | 399 | |||
AK170073.1 | - | 3 | 397 | |||
BC049122.1 | - | 3 | 388 | 6813377 |