Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029847.4 | NP_084123.2 | 1 | 243 | ||
GenBank mRNA | AK013194.2 | - | 1 | 195 | ||
AK019515.1 | - | 1 | 209 | |||
AK031147.1 | - | 1 | 231 | |||
AK032812.1 | - | 1 | 209 | |||
AK033968.1 | - | 1 | 244 | |||
AK035464.1 | - | 1 | 164 | |||
AK039765.1 | - | 1 | 239 | |||
AK046378.1 | - | 1 | 244 | |||
AK083188.1 | - | 1 | 197 | |||
AK084090.1 | - | 1 | 229 | |||
AK086667.1 | - | 1 | 202 | |||
AK144817.1 | - | 1 | 211 | |||
AK165970.1 | - | 1 | 211 | |||
BC046790.1 | - | 1 | 217 | 5717223 | ||
BC058351.1 | - | 1 | 217 | 5717223 |