Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000338.2 | NP_000329.2 | 3 | 725 | ||
NM_001184832.1 | NP_001171761.1 | 3 | 725 | |||
XM_005254606.1 | XP_005254663.1 | 3 | 725 | |||
GenBank mRNA | AK295375.1 | - | 2 | 178 | ||
AK298312.1 | - | 3 | 724 | |||
BC040138.2 | - | 2 | 695 | |||
EF559316.2 | - | 2 | 523 | |||
U58130.1 | - | 2 | 528 |