Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpbp1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029868.2 | NP_084144.2 | 12 | 2538 | ||
GenBank mRNA | AK037224.1 | - | 12 | 2559 | ||
AK077148.1 | - | 12 | 2508 | |||
AK133761.1 | - | 12 | 2554 | |||
BC066034.1 | - | 12 | 2538 | 6836605 |