Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164609.1 | NP_001158081.1 | 3, 4 | 245 | ||
NM_001164610.1 | NP_001158082.1 | 3, 4 | 245 | |||
NM_001164611.1 | NP_001158083.1 | 3, 4 | 245 | |||
NM_029945.3 | NP_084221.2 | 3, 4 | 245 | |||
GenBank mRNA | AK019466.2 | - | 3, 4 | 246 | ||
AK031029.1 | - | 3, 4 | 239 | |||
AK031685.1 | - | 3, 4 | 244 | |||
AK035892.1 | - | 3, 4 | 237 | |||
AK036055.1 | - | 3, 4 | 237 | |||
AK036380.1 | - | 3, 4 | 241 | |||
AK129354.1 | - | 3, 4 | 222 | |||
AK138720.1 | - | 3, 4 | 219 | |||
AK164234.1 | - | 3, 4 | 237 | |||
AK164456.1 | - | 3, 4 | 219 | |||
BC026767.1 | - | 3, 4 | 218 | 5137008 |